Assay Design



Gene Transcripts

Gene Symbol
TEDDM1
Entrez Gene ID
102186237
Gene Name
transmembrane epididymal protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 62112233 - 62113239 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005709588.3 XP_005709645.3 1 - 1 929 74

View Full Product Details