Assay Design



Gene Transcripts

Gene Symbol
CLEC17A
Entrez Gene ID
102186854
Gene Name
C-type lectin domain containing 17A
Gene Aliases
-
Chromosome Location
Chr.7: 96588753 - 96609047 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051679.1 XP_017907168.1 3 - 4 203 90

View Full Product Details