Assay Design



Gene Transcripts

Gene Symbol
GUCA2B
Entrez Gene ID
102180911
Gene Name
guanylate cyclase activator 2B
Gene Aliases
-
Chromosome Location
Chr.3: 16750512 - 16753709 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678616.3 XP_005678673.1 1 - 2 117 68

View Full Product Details