Assay Design



Gene Transcripts

Gene Symbol
KCNG1
Entrez Gene ID
102169395
Gene Name
potassium voltage-gated channel modifier subfamily G member 1
Gene Aliases
-
Chromosome Location
Chr.13: 78433473 - 78450187 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058039.1 XP_017913528.1 2 - 3 1060 102

View Full Product Details