Assay Design



Gene Transcripts

Gene Symbol
C15H11orf16
Entrez Gene ID
102182424
Gene Name
chromosome 15 C11orf16 homolog
Gene Aliases
-
Chromosome Location
Chr.15: 38720315 - 38728682 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059510.1 XP_017914999.1 2 - 3 607 79

View Full Product Details