Assay Design



Gene Transcripts

Gene Symbol
BRK1
Entrez Gene ID
102180975
Gene Name
BRICK1 subunit of SCAR/WAVE actin nucleating complex
Gene Aliases
-
Chromosome Location
Chr.22: 16552010 - 16559675 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695612.3 XP_005695669.1 1 - 2 249 73
GenBank mRNA JO363185.2 - 2 - 3 998 73
KA323094.1 - 1 - 2 195 73

View Full Product Details