Assay Design



Gene Transcripts

Gene Symbol
ZCCHC24
Entrez Gene ID
102172513
Gene Name
zinc finger CCHC-type containing 24
Gene Aliases
-
Chromosome Location
Chr.28: 10599599 - 10665512 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042253.1 XP_017897742.1 3 - 4 612 62
GenBank mRNA JO646010.2 - 3 - 4 638 62

View Full Product Details