Assay Design



Gene Transcripts

Gene Symbol
BEST4
Entrez Gene ID
102178504
Gene Name
bestrophin 4
Gene Aliases
-
Chromosome Location
Chr.3: 19533281 - 19537922 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043437.1 XP_017898926.1 5 - 6 715 90

View Full Product Details