Assay Design



Gene Transcripts

Gene Symbol
FAM32A
Entrez Gene ID
106502278
Gene Name
family with sequence similarity 32 member A
Gene Aliases
-
Chromosome Location
Chr.7: 101929611 - 101935724 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013965028.2 XP_013820482.1 1 - 2 155 81

View Full Product Details