Assay Design



Gene Transcripts

Gene Symbol
FCGBP
Entrez Gene ID
102185859
Gene Name
Fc fragment of IgG binding protein
Gene Aliases
-
Chromosome Location
Chr.18: 50298526 - 50338913 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062404.1 XP_017917893.1 6 - 7 2849 65
XM_018062405.1 XP_017917894.1 5 - 6 1601 65

View Full Product Details