Assay Design



Gene Transcripts

Gene Symbol
ATP6AP2
Entrez Gene ID
102173826
Gene Name
ATPase H+ transporting accessory protein 2
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 30266544 - 30293294 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043923.1 XP_017899412.1 2 - 3 305 87

View Full Product Details