Assay Design



Gene Transcripts

Gene Symbol
DOK3
Entrez Gene ID
102179280
Gene Name
docking protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 71609049 - 71614107 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050596.1 XP_017906085.1 1 - 2 144 68
XM_018050597.1 XP_017906086.1 1 - 2 193 68
GenBank mRNA JT639585.1 - 5 - 6 1300 68

View Full Product Details