Assay Design



Gene Transcripts

Gene Symbol
SERPINB12
Entrez Gene ID
102178964
Gene Name
serpin family B member 12
Gene Aliases
SERPINB13
Chromosome Location
Chr.24: 61967594 - 62007707 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039369.1 XP_017894858.1 11 - 12 1699 70

View Full Product Details