Assay Design



Gene Transcripts

Gene Symbol
TYSND1
Entrez Gene ID
102182668
Gene Name
trypsin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.28: 19284040 - 19294823 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042339.1 XP_017897828.1 3 - 4 2155 99
GenBank mRNA JO408951.2 - 1 - 2 966 99
KA336753.1 - 3 - 4 1645 99

View Full Product Details