Assay Design
Gene Transcripts
- Gene Symbol
- PTTG1
- Entrez Gene ID
-
102183062
- Gene Name
- PTTG1 regulator of sister chromatid separation, securin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 38578338 - 38586073 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO576549.1 |
- |
2 - 3 |
229 |
81 |
JT707332.1 |
- |
1 - 2 |
64 |
81 |
View Full Product Details