Assay Design



Gene Transcripts

Gene Symbol
CHRNA9
Entrez Gene ID
102175104
Gene Name
cholinergic receptor nicotinic alpha 9 subunit
Gene Aliases
-
Chromosome Location
Chr.6: 59906874 - 59920104 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681535.2 XP_005681592.1 4 - 5 974 67

View Full Product Details