Assay Design



Gene Transcripts

Gene Symbol
TGOLN2
Entrez Gene ID
102172070
Gene Name
trans-golgi network protein 2
Gene Aliases
-
Chromosome Location
Chr.11: 49179389 - 49187398 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686729.3 XP_005686786.3 2 - 3 1106 80
GenBank mRNA JO419784.2 - 3 - 4 883 80
JO424341.1 - 3 - 4 936 80

View Full Product Details