Assay Design
Gene Transcripts
- Gene Symbol
- LOC102174762
- Entrez Gene ID
-
102174762
- Gene Name
- carbonic anhydrase 2
- Gene Aliases
-
CA2
- Chromosome Location
-
Chr.14: 5152874 - 5170158 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO612734.2 |
- |
2 - 3 |
582 |
78 |
View Full Product Details