Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018046412.1 | XP_017901901.1 | 2 - 3 | 134 | 82 |
XM_018046413.1 | XP_017901902.1 | 2 - 3 | 285 | 82 | |
GenBank mRNA | JO644036.2 | - | 1 - 2 | 1111 | 82 |
KA345529.1 | - | 2 - 3 | 313 | 82 |