Assay Design



Gene Transcripts

Gene Symbol
SPIN4
Entrez Gene ID
102180204
Gene Name
spindlin family member 4
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 57105227 - 57108382 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044058.1 XP_017899547.1 1 - 1 1105 103

View Full Product Details