Assay Design



Gene Transcripts

Gene Symbol
TAFA3
Entrez Gene ID
102172780
Gene Name
TAFA chemokine like family member 3
Gene Aliases
FAM19A3
Chromosome Location
Chr.3: 89984670 - 89994200 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013962526.2 XP_013817980.2 5 - 6 757 78
XM_018045926.1 XP_017901415.1 6 - 7 712 78

View Full Product Details