Assay Design



Gene Transcripts

Gene Symbol
TNIP2
Entrez Gene ID
102184610
Gene Name
TNFAIP3 interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.6: 116553471 - 116579886 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049837.1 XP_017905326.1 5 - 6 1152 88
GenBank mRNA JO630007.2 - 5 - 6 1150 88
KA331611.1 - 1 - 2 714 88

View Full Product Details