Assay Design



Gene Transcripts

Gene Symbol
PIRT
Entrez Gene ID
102174973
Gene Name
phosphoinositide interacting regulator of transient receptor potential channels
Gene Aliases
-
Chromosome Location
Chr.19: 29486017 - 29496931 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064003.1 XP_017919492.1 2 - 2 381 74

View Full Product Details