Assay Design



Gene Transcripts

Gene Symbol
IFI6
Entrez Gene ID
102185230
Gene Name
interferon alpha inducible protein 6
Gene Aliases
-
Chromosome Location
Chr.2: 10302851 - 10306586 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056827.1 XP_017912316.1 4 - 5 441 90
XM_018056828.1 XP_017912317.1 5 - 6 509 90
GenBank mRNA JO632972.2 - 1 - 2 253 90
JR129141.1 - - 2057 90

View Full Product Details