Assay Design



Gene Transcripts

Gene Symbol
NINJ1
Entrez Gene ID
100861274
Gene Name
ninjurin 1
Gene Aliases
-
Chromosome Location
Chr.8: 84259734 - 84271046 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052500.1 XP_017907989.1 4 - 4 862 97
GenBank mRNA JO439614.2 - 1 - 1 307 97

View Full Product Details