Assay Design



Gene Transcripts

Gene Symbol
Ahsg
Entrez Gene ID
100135479
Gene Name
alpha 2-HS glycoprotein
Gene Aliases
FETUA
Chromosome Location
Chr.Un|NT_176412: 51622730 - 51630327 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172913.1 NP_001166384.1 2 - 3 351 65
GenBank mRNA AB006443.1 - 2 - 3 351 65

View Full Product Details