Assay Design



Gene Transcripts

Gene Symbol
Ptgr1
Entrez Gene ID
100135572
Gene Name
prostaglandin reductase 1
Gene Aliases
Ltb4dh
Chromosome Location
Chr.Un|NT_176326: 3649185 - 3672490 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172980.1 NP_001166451.1 7 - 8 803 86
GenBank mRNA AB021219.1 - 7 - 8 803 86

View Full Product Details