Assay Design



Gene Transcripts

Gene Symbol
Cxcl1
Entrez Gene ID
100135510
Gene Name
chemokine (C-X-C motif) ligand 1
Gene Aliases
GRO, SCYB1
Chromosome Location
Chr.Un|NT_176418: 27585173 - 27587105 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172938.1 NP_001166409.1 3 - 4 300 84
GenBank mRNA AF052004.1 - 3 - 4 300 84
U95809.1 - 3 - 4 300 84
U95810.1 - 3 - 4 300 84

View Full Product Details