Assay Design



Gene Transcripts

Gene Symbol
Opn1mw
Entrez Gene ID
100135586
Gene Name
opsin 1 (cone pigments), medium-wave-sensitive (color blindness, deutan)
Gene Aliases
GCP
Chromosome Location
Chr.Un|NT_176271: 1653382 - 1660256 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172989.1 NP_001166460.1 2 - 3 421 80
GenBank mRNA AF031529.1 - 1 - 2 80
AF132042.1 - 2 - 3 421 80

View Full Product Details