Assay Design



Gene Transcripts

Gene Symbol
Hsd11b1
Entrez Gene ID
100135542
Gene Name
hydroxysteroid 11-beta dehydrogenase 1
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176406: 14260175 - 14281260 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013157260.2 XP_013012714.1 4 - 5 408 71
GenBank mRNA AY535424.1 - 3 - 4 328 71

View Full Product Details