Assay Design



Gene Transcripts

Gene Symbol
Gnai1
Entrez Gene ID
100135528
Gene Name
G protein subunit alpha i1
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176388: 19733791 - 19761257 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172953.1 NP_001166424.1 1 - 2 159 76
GenBank mRNA D21232.1 - 1 - 2 159 76

View Full Product Details