Assay Design



Gene Transcripts

Gene Symbol
Oprl1
Entrez Gene ID
100135588
Gene Name
opioid related nociceptin receptor 1
Gene Aliases
OOR
Chromosome Location
Chr.Un|NT_176300: 272606 - 279793 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172991.1 NP_001166462.1 1 - 2 276 88
XM_005000360.3 XP_005000417.1 2 - 3 350 88
XM_013144564.2 XP_013000018.1 2 - 3 280 88
GenBank mRNA U04369.1 - 1 - 2 276 88

View Full Product Details