Assay Design



Gene Transcripts

Gene Symbol
Ccl11
Entrez Gene ID
100135493
Gene Name
C-C motif chemokine ligand 11
Gene Aliases
SCYA11
Chromosome Location
Chr.Un|NT_176387: 12640305 - 12642559 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172824.1 NP_001166295.1 1 - 2 142 71
GenBank mRNA U18941.1 - 1 - 2 142 71
X77603.1 - 1 - 2 114 71

View Full Product Details