Assay Design



Gene Transcripts

Gene Symbol
Guca2b
Entrez Gene ID
100135535
Gene Name
guanylate cyclase activator 2B
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176239: 1424919 - 1427105 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172958.1 NP_001166429.1 2 - 3 318 59
GenBank mRNA Z74738.1 - 2 - 3 318 59

View Full Product Details