Assay Design



Gene Transcripts

Gene Symbol
Nme1
Entrez Gene ID
100379567
Gene Name
NME/NM23 nucleoside diphosphate kinase 1
Gene Aliases
NDPK-A
Chromosome Location
Chr.Un|NT_176366: 6817708 - 6822870 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172873.1 NP_001166344.1 2 - 3 238 75
GenBank mRNA AY017306.1 - 2 - 3 238 75

View Full Product Details