Assay Design



Gene Transcripts

Gene Symbol
Eno1
Entrez Gene ID
100306968
Gene Name
enolase 1
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176394: 24427678 - 24448149 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173073.1 NP_001166544.1 8 - 9 1080 80
XM_005004205.3 XP_005004262.1 9 - 10 1209 80
XM_005004206.3 XP_005004263.1 9 - 10 1169 80
XM_005004207.3 XP_005004264.1 9 - 10 1209 80
GenBank mRNA EU306867.1 - 8 - 9 1080 80

View Full Product Details