Assay Design



Gene Transcripts

Gene Symbol
Bri3
Entrez Gene ID
100379532
Gene Name
brain protein I3
Gene Aliases
I3
Chromosome Location
Chr.Un|NT_176389: 6378509 - 6386434 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172827.1 NP_001166298.1 2 - 3 247 83
GenBank mRNA AB058506.1 - 2 - 3 247 83

View Full Product Details