Assay Design



Gene Transcripts

Gene Symbol
Edem2
Entrez Gene ID
34714
Gene Name
ER degradation enhancer, mannosidase alpha-like 2
Gene Aliases
CG5682, Dmel\CG5682, Dmel_CG5682, EDEM2, dEDEM2, edem2
Chromosome Location
Chr.2L: 13192201 - 13197281 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032480
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273505.1 NP_001260434.1 3 - 4 969 57
NM_135767.2 NP_609611.1 3 - 4 969 57
GenBank mRNA BT056246.1 - 3 - 4 966 57

View Full Product Details