Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_135969.4 | NP_609813.1 | 9 - 10 | 1610 | 74 |
NM_165194.3 | NP_724016.1 | 9 - 10 | 1537 | 74 | |
NM_165195.3 | NP_724017.1 | 9 - 10 | 1857 | 74 | |
NM_206002.2 | NP_995724.2 | 9 - 10 | 1829 | 74 | |
GenBank mRNA | AY051835.1 | - | 9 - 10 | 1576 | 74 |