Assay Design
Gene Transcripts
- Gene Symbol
- Tsp47F
- Entrez Gene ID
-
36230
- Gene Name
- Tetraspanin 47F
- Gene Aliases
-
CG9033, Dm.Tsp47F, Dmel\CG9033, Dmel_CG9033
- Chromosome Location
-
Chr.2R: 11377193 - 11379177 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033629
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165828.3 |
NP_725044.1 |
2 - 3 |
406 |
70 |
View Full Product Details