Assay Design



Gene Transcripts

Gene Symbol
muskelin
Entrez Gene ID
36386
Gene Name
CG8811-PA
Gene Aliases
CG8811, DmMK, Dmel\CG8811, Dmel_CG8811
Chromosome Location
Chr.2R: 12628697 - 12632378 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033757
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273992.1 NP_001260921.1 7 - 8 1776 114
NM_136957.3 NP_610801.1 6 - 7 1839 114
NM_165928.1 NP_725200.1 7 - 8 1873 114
GenBank mRNA BT150328.1 - 7 - 8 1872 114

View Full Product Details