Assay Design
Gene Transcripts
- Gene Symbol
- Ccn
- Entrez Gene ID
-
39972
- Gene Name
- CG32183-PB
- Gene Aliases
-
CG13735, CG13736, CG32183, Dmel\CG32183, Dmel_CG32183, dCCN
- Chromosome Location
-
Chr.3L: 17690015 - 17748839 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052183
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168744.2 |
NP_730294.1 |
3 - 4 |
646 |
65 |
GenBank mRNA |
BT011496.1 |
- |
3 - 4 |
648 |
65 |
View Full Product Details