Assay Design



Gene Transcripts

Gene Symbol
verm
Entrez Gene ID
40149
Gene Name
vermiform
Gene Aliases
CG8756, DmCDA2, Dmel\CG8756, Dmel_CG8756, LCBP1, N63, Verm, hlm, l(3)76BDx
Chromosome Location
Chr.3L: 19690033 - 19695260 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261341
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169998.3 NP_001163469.1 3 - 4 245 57

View Full Product Details