Assay Design



Gene Transcripts

Gene Symbol
sick
Entrez Gene ID
35277
Gene Name
sickie
Gene Aliases
CG10662, CG13958, CG13959, CG13960, CG13961, CG13964, CG33316, CG34343, CG42589, CG43720, Dm_2L:104545, Dmel\CG43720, Dmel_CG43720
Chromosome Location
Chr.2L: 19796365 - 19958424 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263873
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103713.4 NP_001097183.1 5 - 6 2115 60
NM_001273674.1 NP_001260603.1 7 - 8 2978 60
NM_001273676.1 NP_001260605.1 5 - 6 2231 60
NM_001273677.1 NP_001260606.1 6 - 7 3997 60
NM_001273678.1 NP_001260607.1 6 - 7 4683 60
NM_001273680.2 NP_001260609.1 7 - 8 2883 60
NM_001273681.1 NP_001260610.1 5 - 6 2436 60
NM_206007.5 NP_995729.2 7 - 8 2883 60
NM_206008.4 NP_995730.2 7 - 8 4850 60

View Full Product Details