Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_136429.4 | NP_610273.1 | 3 - 4 | 1032 | 103 |
NM_165518.3 | NP_724547.1 | 3 - 4 | 1634 | 103 | |
NM_165519.3 | NP_724548.1 | 2 - 3 | 1711 | 103 | |
NM_165521.3 | NP_724550.1 | 3 - 4 | 580 | 103 | |
GenBank mRNA | BT001503.1 | - | 2 - 3 | 1687 | 103 |
BT001556.1 | - | 3 - 4 | 1021 | 103 | |
BT003263.1 | - | 3 - 4 | 523 | 103 |