Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001273940.1 | NP_001260869.1 | 2 - 3 | 329 | 103 |
NM_078960.5 | NP_523684.2 | 5 - 6 | 974 | 103 | |
NM_165772.4 | NP_724934.1 | 5 - 6 | 856 | 103 | |
NM_165773.3 | NP_724935.1 | 6 - 7 | 911 | 103 | |
NM_176124.3 | NP_788304.1 | 5 - 6 | 783 | 103 | |
NM_176125.3 | NP_788305.1 | 5 - 6 | 838 | 103 | |
NM_176126.3 | NP_788306.1 | 5 - 6 | 756 | 103 | |
NM_176127.3 | NP_788307.1 | 5 - 6 | 866 | 103 | |
NM_206079.2 | NP_995801.1 | 4 - 5 | 986 | 103 | |
NM_206080.2 | NP_995802.1 | 4 - 5 | 738 | 103 | |
GenBank mRNA | AY121631.1 | - | 6 - 7 | 892 | 103 |
BT046134.2 | - | 5 - 6 | 991 | 103 | |
BT124767.1 | - | 2 - 3 | 329 | 103 | |
M29602.1 | - | 4 - 5 | 1751 | 103 | |
M29731.1 | - | 5 - 6 | 760 | 103 | |
M29732.1 | - | 5 - 6 | 1114 | 103 | |
M30151.1 | - | 5 - 6 | 784 | 103 | |
M86660.1 | - | 4 - 5 | 849 | 103 |