Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104026.2 | NP_001097496.1 | 10 - 11 | 5623 | 81 |
NM_001104027.2 | NP_001097497.1 | 11 - 12 | 5923 | 81 | |
NM_001274447.1 | NP_001261376.1 | 11 - 12 | 5938 | 81 | |
NM_001274448.1 | NP_001261377.1 | 10 - 11 | 5480 | 81 | |
NM_001274449.1 | NP_001261378.1 | 10 - 11 | 5566 | 81 | |
NM_080026.3 | NP_524765.2 | 10 - 11 | 5566 | 81 | |
NM_206270.2 | NP_995992.1 | 10 - 11 | 5581 | 81 | |
GenBank mRNA | BT050449.1 | - | 10 - 11 | 5564 | 81 |