Assay Design
Gene Transcripts
- Gene Symbol
- Prosbeta2R1
- Entrez Gene ID
-
31511
- Gene Name
- Proteasome beta2 subunit-related 1
- Gene Aliases
-
CG18341, Dmel\CG18341, Dmel_CG18341, beta2R1
- Chromosome Location
-
Chr.X: 5808954 - 5810158 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0029812
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132039.3 |
NP_572267.1 |
1 - 2 |
144 |
69 |
View Full Product Details