Assay Design



Gene Transcripts

Gene Symbol
Grip
Entrez Gene ID
50391
Gene Name
Glutamate receptor interacting protein
Gene Aliases
CG14447, CG5980, DGrip, Dgrip, Dmel\CG14447, Dmel_CG14447, GRIP, dGRIP, dgrip, grip
Chromosome Location
Chr.X: 5966279 - 5986699 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029830
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297991.1 NP_001284920.1 6 - 7 1156 54
NM_001297992.1 NP_001284921.1 5 - 6 1107 54
NM_132057.3 NP_572285.2 5 - 6 1294 54
GenBank mRNA BT044143.1 - 5 - 6 1293 54

View Full Product Details