Assay Design



Gene Transcripts

Gene Symbol
Grip
Entrez Gene ID
50391
Gene Name
Glutamate receptor interacting protein
Gene Aliases
CG14447, CG5980, DGrip, Dgrip, Dmel\CG14447, Dmel_CG14447, GRIP, dGRIP, dgrip, grip
Chromosome Location
Chr.X: 5966279 - 5986699 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029830
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297991.1 NP_001284920.1 9 - 10 2747 76
NM_001297992.1 NP_001284921.1 8 - 9 2698 76
NM_132057.3 NP_572285.2 8 - 9 2885 76
GenBank mRNA BT044143.1 - 8 - 9 2884 76

View Full Product Details